accessibility menu, dialog, popup

UserName

NH2BrHNH2N

CAS RN 38875-53-5

IUPAC Name: 2,3-diamino-5-bromopyridin-1-ium
Synonyms: 2,3-diamino-5-bromopyridin-1-ium
Molecular Weight (g/mol): 189.04
Molecular Formula: C5H7BrN3
InChi Key: YRGMYJUKFJPNPD-UHFFFAOYSA-O
SMILES: NC1=CC(Br)=C[NH+]=C1N
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (3)
  • (3)
  • (2)
  • (2)