missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diamino-5-bromopyridine 98.0+%, TCI America™

Catalog No. D29165G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29165G 5 g
D291625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D29165G Supplier TCI America Supplier No. D29165G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 38875-53-5 | C5H7BrN3 | 189.04 g/mol

Chemical Identifiers

CAS 38875-53-5
Molecular Formula C5H7BrN3
Molecular Weight (g/mol) 189.04
MDL Number MFCD00460094
InChI Key YRGMYJUKFJPNPD-UHFFFAOYSA-O
Synonym 2,3-diamino-5-bromopyridine, 5-bromo-2,3-diaminopyridine, 5-bromo-2,3-pyridinediamine, 2,3-pyridinediamine, 5-bromo, 3-amino-5-bromopyridin-2-yl amine, abbypharma ap-17-5972, 5-brompyridin-2,3-diamin, pubchem5383, acmc-1adjl, 5-brompyridin-2,3-diamine
PubChem CID 691156
IUPAC Name 2,3-diamino-5-bromopyridin-1-ium
SMILES NC1=CC(Br)=C[NH+]=C1N

Specifications

Melting Point 161°C
Quantity 5 g
Formula Weight 188.03
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Diamino-5-bromopyridine