missing translation for 'onlineSavingsMsg'
Learn More

2,3-Diamino-5-bromo-6-methylpyridine 98.0+%, TCI America™

Catalog No. p-7128393
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46581G 1 g
D46585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46581G Supplier TCI America Supplier No. D46581G

Chemical Identifiers

CAS 59352-90-8
Molecular Formula C6H8BrN3
Molecular Weight (g/mol) 202.055
MDL Number MFCD00160167
InChI Key XFHAYQGHHKCVBA-UHFFFAOYSA-N
Synonym 5-bromo-6-methyl-2,3-pyridinediamine, 2,3-diamino-5-bromo-6-picoline, 2,3-pyridinediamine, 5-bromo-6-methyl, 2,3-diamino-5-bromo-6-methylpyridine, 5-bromo-2,3-diamino-6-methylpyridine, 5-bromo-6-methyl-pyridine-2,3-diamine, 2,3-diamino-5-bromo-6-methylpyridine 2,3-diamino-5-bromo-6-picoline, 5,6-diamino-3-bromo-, a-picoline, pubchem5382, 3-bromo-5,6-diamino-2-picoline
PubChem CID 101022
IUPAC Name 5-bromo-6-methylpyridine-2,3-diamine
SMILES CC1=C(C=C(C(=N1)N)N)Br

Specifications

Melting Point 136°C
Quantity 1 g
Formula Weight 202.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Diamino-5-bromo-6-methylpyridine