accessibility menu, dialog, popup

UserName

CAS RN 59352-90-8

IUPAC Name: 5-bromo-6-methylpyridine-2,3-diamine
Synonyms: 5-bromo-6-methylpyridine-2,3-diamine
Molecular Weight (g/mol): 202.06
Molecular Formula: C6H8BrN3
InChi Key: XFHAYQGHHKCVBA-UHFFFAOYSA-N
SMILES: CC1=NC(N)=C(N)C=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results