missing translation for 'onlineSavingsMsg'
Learn More

2,3-Di(p-tolyl)-5-phenyltetrazolium Chloride 98.0+%, TCI America™

Catalog No. B0345100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0345100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B0345100MG Supplier TCI America Supplier No. B0345100MG
Only null left

Chemical Identifiers

CAS 104497-77-0
Molecular Formula C21H19ClN4
Molecular Weight (g/mol) 362.86
MDL Number MFCD00060002
InChI Key OUFYUKONKRMAMS-UHFFFAOYSA-M

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 362.86
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Di(p-tolyl)-5-phenyltetrazolium Chloride
EINECSNumber (5)-5949
TSCA No