CAS RN 104497-77-0
IUPAC Name:
2,3-bis(4-methylphenyl)-5-phenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Synonyms:
2,3-bis(4-methylphenyl)-5-phenyl-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Molecular Weight (g/mol):
362.86
Molecular Formula:
C21H19ClN4
InChi Key:
OUFYUKONKRMAMS-UHFFFAOYSA-M
SMILES:
[Cl-].CC1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=C(C)C=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
2,3-Di(p-tolyl)-5-phenyltetrazolium Chloride 98.0+%, TCI America™
CAS: 104497-77-0 Molecular Formula: C21H19ClN4 Molecular Weight (g/mol): 362.86 MDL Number: MFCD00060002 InChI Key: OUFYUKONKRMAMS-UHFFFAOYSA-M