missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Chloropropoxy)tetrahydro-2H-pyran 97.0+%, TCI America™

Catalog No. C207225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C207225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C207225G Supplier TCI America Supplier No. C207225G
Only null left

Chemical Identifiers

CAS 42330-88-1
Molecular Formula C8H15ClO2
Molecular Weight (g/mol) 178.656
MDL Number MFCD00023846
InChI Key QFXXARKSLAKVRL-UHFFFAOYSA-N
PubChem CID 254954
IUPAC Name 2-(3-chloropropoxy)oxane
SMILES C1CCOC(C1)OCCCCl

Specifications

Color Colorless
Boiling Point 107°C
Quantity 25 g
Formula Weight 178.66
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(3-Chloropropoxy)tetrahydro-2H-pyran