accessibility menu, dialog, popup

UserName

ClOO

CAS RN 42330-88-1

IUPAC Name: 2-(3-chloropropoxy)oxane
Synonyms: 2-(3-chloropropoxy)oxane
Molecular Weight (g/mol): 178.66
Molecular Formula: C8H15ClO2
InChi Key: QFXXARKSLAKVRL-UHFFFAOYNA-N
SMILES: ClCCCOC1CCCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results