missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Chlorophenyl)ethylamine 98.0+%, TCI America™

Catalog No. p-7125744
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C29555G 5 g
C295525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C29555G Supplier TCI America Supplier No. C29555G
Only null left

Chemical Identifiers

CAS 13078-79-0
Molecular Formula C8H10ClN
Molecular Weight (g/mol) 155.625
MDL Number MFCD00047957
InChI Key NRHVNPYOTNGECT-UHFFFAOYSA-N
Synonym 2-3-chlorophenyl ethylamine, 2-3-chlorophenyl ethanamine, 3-chlorophenethylamine, 2-3-chlorophenyl ethan-1-amine, 2-3-chloro-phenyl-ethylamine, 3-chlorophenylethanamine, 3'-chlorophenethylamine, m-chlorophenethylamine, 2-m-chlorophenyl ethylamine, benzeneethanamine, 3-chloro
PubChem CID 139381
IUPAC Name 2-(3-chlorophenyl)ethanamine
SMILES C1=CC(=CC(=C1)Cl)CCN

Specifications

Color Yellow
Boiling Point 145°C
Quantity 5 g
Formula Weight 155.63
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-(3-Chlorophenyl)ethylamine