accessibility menu, dialog, popup

UserName

CAS RN 13078-79-0

IUPAC Name: 2-(3-chlorophenyl)ethan-1-aminium
Synonyms: 2-(3-chlorophenyl)ethanaminium
Molecular Weight (g/mol): 156.63
Molecular Formula: C8H11ClN
InChi Key: NRHVNPYOTNGECT-UHFFFAOYSA-O
SMILES: [NH3+]CCC1=CC=CC(Cl)=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)