missing translation for 'onlineSavingsMsg'
Learn More

2,3-Butanedithiol 98.0+%, TCI America™

Catalog No. p-7119827
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28885G 5 g
B288825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B28885G Supplier TCI America Supplier No. B28885G
Only null left

Chemical Identifiers

CAS 4532-64-3
Molecular Formula C4H10S2
Molecular Weight (g/mol) 122.244
MDL Number MFCD00010024
InChI Key TWWSEEHCVDRRRI-UHFFFAOYSA-N
Synonym 2,3-Dimercaptobutane
PubChem CID 548353
IUPAC Name butane-2,3-dithiol
SMILES CC(C(C)S)S

Specifications

Color Yellow
Boiling Point 87°C
Quantity 5 g
UN Number 3336
Formula Weight 122.24
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Butanedithiol