accessibility menu, dialog, popup

UserName

H3CSHCH3HS

CAS RN 4532-64-3

IUPAC Name: butane-2,3-dithiol
Synonyms: butane-2,3-dithiol
Molecular Weight (g/mol): 122.24
Molecular Formula: C4H10S2
InChi Key: TWWSEEHCVDRRRI-UHFFFAOYNA-N
SMILES: CC(S)C(C)S
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results