missing translation for 'onlineSavingsMsg'
Learn More

2,3-Butanedione monoxime, 98%

Catalog No. p-4510165
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC150371000 100 g Plastic bottle
AC150375000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC150371000 Supplier Thermo Scientific Chemicals Supplier No. 150371000
Only null left

CAS: 57-71-6 | C4H7NO2 | 101.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57-71-6
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.11
MDL Number MFCD00002116
InChI Key FSEUPUDHEBLWJY-HWKANZROSA-N
Synonym 2,3-butanedione monoxime, diacetyl monoxime, diacetylmonoxime, biacetyl monoxime, biacetyl monooxime, diacetyl monooxime, diacetylmonooxime, 2-oximino-3-butanone, 3-oximino-2-butanone, 2,3-butanedione-2-monoxime
PubChem CID 6409633
ChEBI CHEBI:4480
IUPAC Name (3E)-3-hydroxyiminobutan-2-one
SMILES CC(=O)C(\C)=N\O

Specifications

Melting Point 74°C to 76°C
Color White
Boiling Point 185°C to 186°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula CH3C(=NOH)COCH3
Quantity 100 g
Beilstein 01,772
Solubility Information (5% in water) Clear to slightly hazy
Formula Weight 101.1
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2, 3-Butanedione monoxime
Show More Show Less
EINECSNumber 200-348-5

RUO – Research Use Only