missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Bromopropoxy)tetrahydro-2H-pyran, 98%, stabilized with potassium carbonate, Thermo Scientific™

Chemical Identifiers

CAS 33821-94-2
Molecular Formula C8H15BrO2
Molecular Weight (g/mol) 223.1
InChI Key HJNHUFQGDJLQRS-UHFFFAOYSA-N
Synonym 2-3-bromopropoxy tetrahydro-2h-pyran, 2-3-bromopropoxy oxane, 2-3-bromopropoxy-tetrahydro-2h-pyran, 2-3-bromopropoxy tetrahydropyran, 1-bromo-3-tetrahydropyranyloxypropane, 2-3-bromo-propoxy-tetrahydro-pyran, 2h-pyran, 2-3-bromopropoxy tetrahydro, acmc-1cjr6, 1-bromo-3-2-oxanyloxy propane
PubChem CID 2777988
IUPAC Name 2-(3-bromopropoxy)oxane
SMILES C1CCOC(C1)OCCCBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC399730010
View Documents
Thermo Scientific Chemicals
399730010
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC399730050
View Documents
Thermo Scientific Chemicals
399730050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

33821-94-2
223.1
2-3-bromopropoxy tetrahydro-2h-pyran, 2-3-bromopropoxy oxane, 2-3-bromopropoxy-tetrahydro-2h-pyran, 2-3-bromopropoxy tetrahydropyran, 1-bromo-3-tetrahydropyranyloxypropane, 2-3-bromo-propoxy-tetrahydro-pyran, 2h-pyran, 2-3-bromopropoxy tetrahydro, acmc-1cjr6, 1-bromo-3-2-oxanyloxy propane
2-(3-bromopropoxy)oxane
C8H15BrO2
HJNHUFQGDJLQRS-UHFFFAOYSA-N
2777988
C1CCOC(C1)OCCCBr

Specifications

33821-94-2
64.5°C (0.8mmHg)
97.5% min. (GC)
C8H15BrO2
1.314
HJNHUFQGDJLQRS-UHFFFAOYSA-N
2-(3-bromopropoxy)oxane
2777988
98%
1.3140g/mL
106°C
Glass bottle
1g
2-3-bromopropoxy tetrahydro-2h-pyran, 2-3-bromopropoxy oxane, 2-3-bromopropoxy-tetrahydro-2h-pyran, 2-3-bromopropoxy tetrahydropyran, 1-bromo-3-tetrahydropyranyloxypropane, 2-3-bromo-propoxy-tetrahydro-pyran, 2h-pyran, 2-3-bromopropoxy tetrahydro, acmc-1cjr6, 1-bromo-3-2-oxanyloxy propane
C1CCOC(C1)OCCCBr
223.1
223.1
2-(3-Bromopropoxy)tetrahydro-2H-pyran, Stabilized with potassium carbonate
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only