missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(3-Bromopropoxy)tetrahydro-2H-pyran, 98%, stabilized with potassium carbonate, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 399730010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
| 2-(3-Bromopropoxy)tetrahydro-2H-pyran | |
| 33821-94-2 | |
| 64.5°C (0.8mmHg) | |
| 97.5% min. (GC) | |
| C8H15BrO2 | |
| 1.314 | |
| HJNHUFQGDJLQRS-UHFFFAOYSA-N | |
| 2-(3-bromopropoxy)oxane | |
| 2777988 | |
| 98% |
| Stabilized with potassium carbonate | |
| 1.3140g/mL | |
| 106°C | |
| Glass bottle | |
| 1g | |
| 2-3-bromopropoxy tetrahydro-2h-pyran, 2-3-bromopropoxy oxane, 2-3-bromopropoxy-tetrahydro-2h-pyran, 2-3-bromopropoxy tetrahydropyran, 1-bromo-3-tetrahydropyranyloxypropane, 2-3-bromo-propoxy-tetrahydro-pyran, 2h-pyran, 2-3-bromopropoxy tetrahydro, acmc-1cjr6, 1-bromo-3-2-oxanyloxy propane | |
| C1CCOC(C1)OCCCBr | |
| 223.1 | |
| 223.1 |
Chemical Identifiers
| 33821-94-2 | |
| 223.1 | |
| 2-3-bromopropoxy tetrahydro-2h-pyran, 2-3-bromopropoxy oxane, 2-3-bromopropoxy-tetrahydro-2h-pyran, 2-3-bromopropoxy tetrahydropyran, 1-bromo-3-tetrahydropyranyloxypropane, 2-3-bromo-propoxy-tetrahydro-pyran, 2h-pyran, 2-3-bromopropoxy tetrahydro, acmc-1cjr6, 1-bromo-3-2-oxanyloxy propane | |
| 2-(3-bromopropoxy)oxane |
| C8H15BrO2 | |
| HJNHUFQGDJLQRS-UHFFFAOYSA-N | |
| 2777988 | |
| C1CCOC(C1)OCCCBr |
Safety and Handling
GHS Signal Word: Warning
RUO â Research Use Only