missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
| CAS | 4373-60-8 |
|---|---|
| Molecular Formula | C11H8BrN |
| Molecular Weight (g/mol) | 234.1 |
| InChI Key | WLPFTJXVEBANAM-UHFFFAOYSA-N |
| Synonym | 2-3-bromophenyl pyridine, pyridine, 2-3-bromophenyl, 2-3-bromo-phenyl-pyridine, 2-3'-bromophenyl pyridine, 3-2-pyridinyl phenyl bromide, 3,5-pyrid-2-yl bromobenzene |
| PubChem CID | 11115506 |
| IUPAC Name | 2-(3-bromophenyl)pyridine |
| SMILES | C1=CC=NC(=C1)C2=CC(=CC=C2)Br |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC457560050
|
N/A | 5g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 4373-60-8 | |
| 234.1 | |
| 2-3-bromophenyl pyridine, pyridine, 2-3-bromophenyl, 2-3-bromo-phenyl-pyridine, 2-3'-bromophenyl pyridine, 3-2-pyridinyl phenyl bromide, 3,5-pyrid-2-yl bromobenzene | |
| 2-(3-bromophenyl)pyridine |
| C11H8BrN | |
| WLPFTJXVEBANAM-UHFFFAOYSA-N | |
| 11115506 | |
| C1=CC=NC(=C1)C2=CC(=CC=C2)Br |
Specifications
| 4373-60-8 | |
| 96% min. (GC) | |
| 2-3-bromophenyl pyridine, pyridine, 2-3-bromophenyl, 2-3-bromo-phenyl-pyridine, 2-3'-bromophenyl pyridine, 3-2-pyridinyl phenyl bromide, 3,5-pyrid-2-yl bromobenzene | |
| C1=CC=NC(=C1)C2=CC(=CC=C2)Br | |
| 234.1 | |
| 234.1 | |
| 2-(3-Bromophenyl)pyridine |
| Authentic | |
| C11H8BrN | |
| WLPFTJXVEBANAM-UHFFFAOYSA-N | |
| 2-(3-bromophenyl)pyridine | |
| 11115506 | |
| 97% |
Safety and Handling
GHS H Statement Causes skin irritation. Causes serious eye irritation.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
GHS Signal Word: Warning
RUO – Research Use Only