missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(3-Bromophenyl)propan-2-ol, 97%, Thermo Scientific™
Specifications
Specifications
| Boiling Point | 100°C (3.0 Torr) |
| Assay Percent Range | 96% min. (GC) |
| Packaging | Glass Bottle |
| Quantity | 1g |
| Formula Weight | 215.09 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-(3-Bromophenyl)propan-2-ol |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF |
RUO – Research Use Only