accessibility menu, dialog, popup

UserName

CH3H3CBrOH

CAS RN 30951-66-7

IUPAC Name: 2-(3-bromophenyl)propan-2-ol
Synonyms: 2-(3-bromophenyl)propan-2-ol
Molecular Weight (g/mol): 215.09
Molecular Formula: C9H11BrO
InChi Key: ZRFMJMFYMQAUDO-UHFFFAOYSA-N
SMILES: CC(C)(O)C1=CC(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)