missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(3-Bromophenyl)ethyl Alcohol 98.0+%, TCI America™
$126.76 - $622.59
Chemical Identifiers
| CAS | 28229-69-8 |
|---|---|
| Molecular Formula | C8H9BrO |
| Molecular Weight (g/mol) | 201.06 |
| MDL Number | MFCD00191857 |
| InChI Key | PTTFLKHCSZSFOL-UHFFFAOYSA-N |
| Synonym | 3-bromophenethyl alcohol, 2-3-bromophenyl ethanol, 2-3-bromophenyl ethan-1-ol, benzeneethanol, 3-bromo, 3-bromophenethylalcohol, 3-bromobenzeneethanol, 3-2-hydroxyethyl bromobenzene, 2-3-bromophenyl ethyl alcohol, pubchem14821, 3-bromo-phenethyl-alcohol |
| PubChem CID | 2734090 |
| IUPAC Name | 2-(3-bromophenyl)ethan-1-ol |
| SMILES | OCCC1=CC=CC(Br)=C1 |
Chemical Identifiers
| 28229-69-8 | |
| 201.06 | |
| PTTFLKHCSZSFOL-UHFFFAOYSA-N | |
| 2734090 | |
| OCCC1=CC=CC(Br)=C1 |
| C8H9BrO | |
| MFCD00191857 | |
| 3-bromophenethyl alcohol, 2-3-bromophenyl ethanol, 2-3-bromophenyl ethan-1-ol, benzeneethanol, 3-bromo, 3-bromophenethylalcohol, 3-bromobenzeneethanol, 3-2-hydroxyethyl bromobenzene, 2-3-bromophenyl ethyl alcohol, pubchem14821, 3-bromo-phenethyl-alcohol | |
| 2-(3-bromophenyl)ethan-1-ol |
Specifications
| 28229-69-8 | |
| 110°C | |
| MFCD00191857 | |
| 3-bromophenethyl alcohol, 2-3-bromophenyl ethanol, 2-3-bromophenyl ethan-1-ol, benzeneethanol, 3-bromo, 3-bromophenethylalcohol, 3-bromobenzeneethanol, 3-2-hydroxyethyl bromobenzene, 2-3-bromophenyl ethyl alcohol, pubchem14821, 3-bromo-phenethyl-alcohol | |
| OCCC1=CC=CC(Br)=C1 | |
| 201.06 | |
| 201.06 | |
| Liquid |
| Yellow | |
| C8H9BrO | |
| 5 g | |
| PTTFLKHCSZSFOL-UHFFFAOYSA-N | |
| 2-(3-bromophenyl)ethan-1-ol | |
| 2734090 | |
| ≥98.0% (GC) | |
| 2-(3-Bromophenyl)ethyl Alcohol |
Safety and Handling
TSCA : No