missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Bromophenyl)ethyl Alcohol 98.0+%, TCI America™

Catalog No. B35175G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35175G 5 g
B351725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35175G Supplier TCI America Supplier No. B35175G
Only null left

Chemical Identifiers

CAS 28229-69-8
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.06
MDL Number MFCD00191857
InChI Key PTTFLKHCSZSFOL-UHFFFAOYSA-N
Synonym 3-bromophenethyl alcohol, 2-3-bromophenyl ethanol, 2-3-bromophenyl ethan-1-ol, benzeneethanol, 3-bromo, 3-bromophenethylalcohol, 3-bromobenzeneethanol, 3-2-hydroxyethyl bromobenzene, 2-3-bromophenyl ethyl alcohol, pubchem14821, 3-bromo-phenethyl-alcohol
PubChem CID 2734090
IUPAC Name 2-(3-bromophenyl)ethan-1-ol
SMILES OCCC1=CC=CC(Br)=C1

Specifications

Color Yellow
Boiling Point 110°C
Quantity 5 g
Formula Weight 201.06
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(3-Bromophenyl)ethyl Alcohol