missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(3-Bromophenyl)-1,3-dioxolane 97.0+%, TCI America™
$103.97 - $421.61
Chemical Identifiers
| CAS | 17789-14-9 |
|---|---|
| Molecular Formula | C9H9BrO2 |
| Molecular Weight (g/mol) | 229.073 |
| MDL Number | MFCD00003209 |
| InChI Key | VYPYKCPWNPPBBX-UHFFFAOYSA-N |
| Synonym | 2-3-bromophenyl-1,3-dioxolane, 3-bromobenzaldehyde ethylene acetal, 1,3-dioxolane, 2-3-bromophenyl, 1-bromo-3-1,3-dioxolan-2-yl benzene, 2-m-bromophenyl-1,3-dioxolane, m-bromobenzaldehyde ethylene acetal, pubchem3730, 2-3-bromophenyl dioxolane, acmc-1c8q7, ksc496m7r |
| PubChem CID | 87306 |
| IUPAC Name | 2-(3-bromophenyl)-1,3-dioxolane |
| SMILES | C1COC(O1)C2=CC(=CC=C2)Br |
Chemical Identifiers
| 17789-14-9 | |
| 229.073 | |
| VYPYKCPWNPPBBX-UHFFFAOYSA-N | |
| 87306 | |
| C1COC(O1)C2=CC(=CC=C2)Br |
| C9H9BrO2 | |
| MFCD00003209 | |
| 2-3-bromophenyl-1,3-dioxolane, 3-bromobenzaldehyde ethylene acetal, 1,3-dioxolane, 2-3-bromophenyl, 1-bromo-3-1,3-dioxolan-2-yl benzene, 2-m-bromophenyl-1,3-dioxolane, m-bromobenzaldehyde ethylene acetal, pubchem3730, 2-3-bromophenyl dioxolane, acmc-1c8q7, ksc496m7r | |
| 2-(3-bromophenyl)-1,3-dioxolane |
Specifications
| 17789-14-9 | |
| C9H9BrO2 | |
| 5 g | |
| VYPYKCPWNPPBBX-UHFFFAOYSA-N | |
| 2-(3-bromophenyl)-1,3-dioxolane | |
| 87306 | |
| ≥97.0% (GC) | |
| 2-(3-Bromophenyl)-1,3-dioxolane |
| Yellow | |
| MFCD00003209 | |
| 2-3-bromophenyl-1,3-dioxolane, 3-bromobenzaldehyde ethylene acetal, 1,3-dioxolane, 2-3-bromophenyl, 1-bromo-3-1,3-dioxolan-2-yl benzene, 2-m-bromophenyl-1,3-dioxolane, m-bromobenzaldehyde ethylene acetal, pubchem3730, 2-3-bromophenyl dioxolane, acmc-1c8q7, ksc496m7r | |
| C1COC(O1)C2=CC(=CC=C2)Br | |
| 229.073 | |
| 229.07 | |
| Liquid |
Safety and Handling
TSCA : No