missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Bromophenyl)-1,3-dioxolane 97.0+%, TCI America™

Catalog No. B20985G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B20985G 5 g
B209825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B20985G Supplier TCI America Supplier No. B20985G

Chemical Identifiers

CAS 17789-14-9
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00003209
InChI Key VYPYKCPWNPPBBX-UHFFFAOYSA-N
Synonym 2-3-bromophenyl-1,3-dioxolane, 3-bromobenzaldehyde ethylene acetal, 1,3-dioxolane, 2-3-bromophenyl, 1-bromo-3-1,3-dioxolan-2-yl benzene, 2-m-bromophenyl-1,3-dioxolane, m-bromobenzaldehyde ethylene acetal, pubchem3730, 2-3-bromophenyl dioxolane, acmc-1c8q7, ksc496m7r
PubChem CID 87306
IUPAC Name 2-(3-bromophenyl)-1,3-dioxolane
SMILES C1COC(O1)C2=CC(=CC=C2)Br

Specifications

Color Yellow
Quantity 5 g
Formula Weight 229.07
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(3-Bromophenyl)-1,3-dioxolane