missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Bromo-4-fluorophenyl)acetonitrile, 96%

Catalog No. AAH6226003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6226003 1 g
AAH6226006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6226003 Supplier Thermo Scientific Chemicals Supplier No. H6226003
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 501420-63-9 | C8H5BrFN | 214.037 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 501420-63-9
Molecular Formula C8H5BrFN
Molecular Weight (g/mol) 214.037
MDL Number MFCD08458154
InChI Key ORKCKZRBHXMWBO-UHFFFAOYSA-N
Synonym 2-3-bromo-4-fluorophenyl acetonitrile, 3-bromo-4-fluorophenylacetonitrile, 3-bromo-4-fluorobenzyl cyanide, benzeneacetonitrile, 3-bromo-4-fluoro, 3-bromo-4-fluorophenyl acetonitrile, 3-bromo-4-fluoro benzyl cyanide, 3-bromo-4-fluorobenzeneacetonitrile, 2-3-bromo-4-fluorophenyl ethanenitrile
PubChem CID 20113897
IUPAC Name 2-(3-bromo-4-fluorophenyl)acetonitrile
SMILES C1=CC(=C(C=C1CC#N)Br)F

Specifications

Quantity 1 g
UN Number UN3439
Formula Weight 214.03
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-(3-Bromo-4-fluorophenyl)acetonitrile
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only