missing translation for 'onlineSavingsMsg'
Learn More

2,3-Bis(4-methoxyphenyl)-5-phenyltetrazolium Chloride 98.0+%, TCI America™

Catalog No. B10561G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B10561G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B10561G Supplier TCI America Supplier No. B10561G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 104497-79-2
Molecular Formula C21H19ClN4O2
Molecular Weight (g/mol) 394.86
MDL Number MFCD00060012
InChI Key BBOUGQFAXQCKHN-UHFFFAOYSA-M

Specifications

Quantity 1 g
Formula Weight 394.86
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Bis(4-methoxyphenyl)-5-phenyltetrazolium Chloride
EINECSNumber (5)-6065
TSCA No