accessibility menu, dialog, popup

UserName

H3CONNNNOH3CCl

CAS RN 104497-79-2

IUPAC Name: 2,3-bis(4-methoxyphenyl)-5-phenyl-3H-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Synonyms: 2,3-bis(4-methoxyphenyl)-5-phenyl-1,2λ⁵,3,4-tetrazol-2-ylium chloride
Molecular Weight (g/mol): 394.86
Molecular Formula: C21H19ClN4O2
InChi Key: BBOUGQFAXQCKHN-UHFFFAOYSA-M
SMILES: [Cl-].COC1=CC=C(C=C1)N1N=C(N=[N+]1C1=CC=C(OC)C=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results