missing translation for 'onlineSavingsMsg'
Learn More

2,3-Bis(4-hydroxyphenyl)propionitrile 98.0+%, TCI America™

Catalog No. B438550MG
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B438550MG 50 mg
B438510MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B438550MG Supplier TCI America Supplier No. B438550MG
Only null left

Chemical Identifiers

CAS 1428-67-7
Molecular Formula C15H13NO2
Molecular Weight (g/mol) 239.274
MDL Number MFCD01695412
InChI Key GHZHWDWADLAOIQ-UHFFFAOYSA-N
Synonym diarylpropionitrile, 2,3-bis 4-hydroxyphenyl propionitrile, 2,3-bis 4-hydroxyphenyl propanenitrile, 2,3-bis 4-hydroxyphenyl-propionitrile, 2,3-bis p-hydroxyphenyl propionitrile, dpn compound, benzenepropanenitrile,4-hydroxy-a-4-hydroxyphenyl, diarylpropionitril, 2,3-bis p-hydroxyphenyl-propionitrile, propionitrile, 2,3-bis p-hydroxyphenyl
PubChem CID 102614
ChEBI CHEBI:63949
IUPAC Name 2,3-bis(4-hydroxyphenyl)propanenitrile
SMILES C1=CC(=CC=C1CC(C#N)C2=CC=C(C=C2)O)O

Specifications

Melting Point 203°C
Color White-Yellow
Quantity 50 mg
Formula Weight 239.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Bis(4-hydroxyphenyl)propionitrile
RTECSNumber UG0900000
TSCA No
Recommended Storage Freezer