missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Bis(4-bromophenyl)-2-butenedinitrile 98.0+%, TCI America™
$203.17 - $596.49
Chemical Identifiers
| CAS | 315203-26-0 |
|---|---|
| Molecular Formula | C16H8Br2N2 |
| Molecular Weight (g/mol) | 388.06 |
| MDL Number | MFCD15072146 |
| InChI Key | RWHQRUSYDYNSTI-UHFFFAOYSA-N |
| PubChem CID | 69745618 |
| IUPAC Name | bis(4-bromophenyl)but-2-enedinitrile |
| SMILES | BrC1=CC=C(C=C1)C(C#N)=C(C#N)C1=CC=C(Br)C=C1 |
Chemical Identifiers
| 315203-26-0 | |
| 388.06 | |
| RWHQRUSYDYNSTI-UHFFFAOYSA-N | |
| bis(4-bromophenyl)but-2-enedinitrile |
| C16H8Br2N2 | |
| MFCD15072146 | |
| 69745618 | |
| BrC1=CC=C(C=C1)C(C#N)=C(C#N)C1=CC=C(Br)C=C1 |
Specifications
| 315203-26-0 | |
| White-Yellow | |
| MFCD15072146 | |
| 3439 | |
| BrC1=CC=C(C=C1)C(C#N)=C(C#N)C1=CC=C(Br)C=C1 | |
| 388.06 | |
| 388.06 | |
| Crystalline Powder |
| 218°C | |
| C16H8Br2N2 | |
| 1 g | |
| RWHQRUSYDYNSTI-UHFFFAOYSA-N | |
| bis(4-bromophenyl)but-2-enedinitrile | |
| 69745618 | |
| ≥98.0% (GC) | |
| 2,3-Bis(4-bromophenyl)-2-butenedinitrile |
Safety and Handling
TSCA : No