missing translation for 'onlineSavingsMsg'
Learn More

2,3-Bis(2,4,5-trimethyl-3-thienyl)maleic Anhydride 97.0+%, TCI America™

Supplier:  TCI America B1534100MG

 View more versions of this product

Catalog No. B1534100MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,3-Bis(2,4,5-trimethyl-3-thienyl)maleic Anhydride
168°C
MFCD00142789
ANYDHJQJXVIYHM-UHFFFAOYSA-N
bis(2,4,5-trimethylthiophen-3-yl)-2,5-dihydrofuran-2,5-dione
11382417
≥97.0% (T)
112440-47-8
C18H18O3S2
100 mg
CC1=C(C)C(=C(C)S1)C1=C(C(=O)OC1=O)C1=C(C)SC(C)=C1C
346.46
346.46
Crystalline Powder

Chemical Identifiers

112440-47-8
346.46
ANYDHJQJXVIYHM-UHFFFAOYSA-N
bis(2,4,5-trimethylthiophen-3-yl)-2,5-dihydrofuran-2,5-dione
C18H18O3S2
MFCD00142789
11382417
CC1=C(C)C(=C(C)S1)C1=C(C(=O)OC1=O)C1=C(C)SC(C)=C1C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions