accessibility menu, dialog, popup

UserName

CH3SOOOSH3CH3CCH3H3CCH3

CAS RN 112440-47-8

IUPAC Name: bis(2,4,5-trimethylthiophen-3-yl)-2,5-dihydrofuran-2,5-dione
Synonyms: bis(2,4,5-trimethylthiophen-3-yl)furan-2,5-dione
Molecular Weight (g/mol): 346.46
Molecular Formula: C18H18O3S2
InChi Key: ANYDHJQJXVIYHM-UHFFFAOYSA-N
SMILES: CC1=C(C)C(=C(C)S1)C1=C(C(=O)OC1=O)C1=C(C)SC(C)=C1C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results