missing translation for 'onlineSavingsMsg'
Learn More

2,3-Bis(2,4,5-trimethyl-3-thienyl)maleic Anhydride 97.0+%, TCI America™

Catalog No. B15341G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15341G 1 g
B1534100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15341G Supplier TCI America Supplier No. B15341G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112440-47-8
Molecular Formula C18H18O3S2
Molecular Weight (g/mol) 346.46
MDL Number MFCD00142789
InChI Key ANYDHJQJXVIYHM-UHFFFAOYSA-N
PubChem CID 11382417
IUPAC Name bis(2,4,5-trimethylthiophen-3-yl)-2,5-dihydrofuran-2,5-dione
SMILES CC1=C(C)C(=C(C)S1)C1=C(C(=O)OC1=O)C1=C(C)SC(C)=C1C

Specifications

Melting Point 168°C
Quantity 1 g
Formula Weight 346.46
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Bis(2,4,5-trimethyl-3-thienyl)maleic Anhydride