missing translation for 'onlineSavingsMsg'
Learn More

2-(3-Benzoylphenyl)propionitrile 98.0+%, TCI America™

Catalog No. p-7119852
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29395G 5 g
B293925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29395G Supplier TCI America Supplier No. B29395G

Chemical Identifiers

CAS 42872-30-0
Molecular Formula C16H13NO
Molecular Weight (g/mol) 235.29
MDL Number MFCD00044435
InChI Key RGYOCHMZSLUCNP-UHFFFAOYNA-N
Synonym 3-(1-Cyanoethyl)benzophenone, 3-Benzoyl-alpha-methylphenylacetonitrile
PubChem CID 39325
IUPAC Name 2-(3-benzoylphenyl)propanenitrile
SMILES CC(C#N)C1=CC=CC(=C1)C(=O)C1=CC=CC=C1

Specifications

Melting Point 57°C
Color White-Yellow
Quantity 5 g
Formula Weight 235.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(3-Benzoylphenyl)propionitrile
RTECSNumber CY1693000
TSCA No