missing translation for 'onlineSavingsMsg'
Learn More

2,3-Benzofuran 99.0+%, TCI America™

Catalog No. B00605G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B00605G 5 g
B006025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B00605G Supplier TCI America Supplier No. B00605G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 271-89-6
Molecular Formula C8H6O
Molecular Weight (g/mol) 118.14
MDL Number MFCD00005847
InChI Key IANQTJSKSUMEQM-UHFFFAOYSA-N
Synonym benzofuran, 2,3-benzofuran, coumarone, benzo b furan, cumarone, benzofurfuran, 1-oxindene, coumaron, 1-oxidene
PubChem CID 9223
ChEBI CHEBI:35260
IUPAC Name 1-benzofuran
SMILES O1C=CC2=CC=CC=C12

Specifications

Boiling Point 175°C
Quantity 5 g
UN Number 1993
Formula Weight 118.14
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3-Benzofuran
RTECSNumber DF6423800
TSCA Yes