missing translation for 'onlineSavingsMsg'
Learn More

2,3-Benzofuran, 99%

Catalog No. p-3735572
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105480100 10 mL Glass bottle
AC105480500 50 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC105480100 Supplier Thermo Scientific Chemicals Supplier No. 105480100
Only null left

CAS: 271-89-6 | C8H6O | 118.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 271-89-6
Molecular Formula C8H6O
Molecular Weight (g/mol) 118.14
MDL Number MFCD00005847
InChI Key IANQTJSKSUMEQM-UHFFFAOYSA-N
Synonym benzofuran, 2,3-benzofuran, coumarone, benzo b furan, cumarone, benzofurfuran, 1-oxindene, coumaron, 1-oxidene
PubChem CID 9223
ChEBI CHEBI:35260
IUPAC Name 1-benzofuran
SMILES O1C=CC2=CC=CC=C12

Specifications

CAS Min % 99.4
CAS Max % 100.0
Color Brown-Yellow to Yellow
Density 1.0700g/mL
Boiling Point 173°C to 175°C
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 99.4 % min. (GC)
Packaging Glass bottle
Refractive Index 1.5655 to 1.5675
Quantity 10 mL
Beilstein 17, 54
Specific Gravity 1.07
Merck Index 15, 1090
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in aq. alkaline solutions
Formula Weight 118.13
Percent Purity 99.50%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2, 3-Benzofuran
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Suspected of causing cancer.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Use personal protective equipment as required.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-982-6

RUO – Research Use Only