missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Benzofluorene 97.0+%, TCI America™
$138.61 - $808.57
Chemical Identifiers
| CAS | 243-17-4 |
|---|---|
| Molecular Formula | C17H12 |
| Molecular Weight (g/mol) | 216.28 |
| MDL Number | MFCD00003595 |
| InChI Key | HAPOJKSPCGLOOD-UHFFFAOYSA-N |
| Synonym | 2,3-benzofluorene, 11h-benzo b fluorene, benzo b fluorene, unii-gmo75ruy0l, 2,3-benzfluorene, gmo75ruy0l, 11h-benzo b fluorene # |
| PubChem CID | 9201 |
| ChEBI | CHEBI:34566 |
| IUPAC Name | 11H-benzo[b]fluorene |
| SMILES | C1C2=CC=CC=C2C2=C1C=C1C=CC=CC1=C2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B0059100MG
|
TCI America
B0059100MG |
100 mg |
Each for $138.61
|
|
|||||
|
B00591G
|
TCI America
B00591G |
1 g |
Each for $808.57
|
|
|||||
Chemical Identifiers
| 243-17-4 | |
| 216.28 | |
| HAPOJKSPCGLOOD-UHFFFAOYSA-N | |
| 9201 | |
| 11H-benzo[b]fluorene |
| C17H12 | |
| MFCD00003595 | |
| 2,3-benzofluorene, 11h-benzo b fluorene, benzo b fluorene, unii-gmo75ruy0l, 2,3-benzfluorene, gmo75ruy0l, 11h-benzo b fluorene # | |
| CHEBI:34566 | |
| C1C2=CC=CC=C2C2=C1C=C1C=CC=CC1=C2 |
Specifications
| 243-17-4 | |
| White-Yellow | |
| C17H12 | |
| 100 mg | |
| HAPOJKSPCGLOOD-UHFFFAOYSA-N | |
| 11H-benzo[b]fluorene | |
| 9201 | |
| 216.28 | |
| Crystalline Powder |
| 213°C | |
| 402°C | |
| MFCD00003595 | |
| 2,3-benzofluorene, 11h-benzo b fluorene, benzo b fluorene, unii-gmo75ruy0l, 2,3-benzfluorene, gmo75ruy0l, 11h-benzo b fluorene # | |
| C1C2=CC=CC=C2C2=C1C=C1C=CC=CC1=C2 | |
| 216.28 | |
| CHEBI:34566 | |
| ≥97.0% (GC) | |
| 2,3-Benzofluorene |
Safety and Handling
RTECSNumber : DF6383500
TSCA : Yes