missing translation for 'onlineSavingsMsg'
Learn More

2,3-Anthracenedicarboxylic Anhydride 93.0+%, TCI America™

Catalog No. A1834100MG
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1834100MG 100 mg
A1834500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1834100MG Supplier TCI America Supplier No. A1834100MG
Only null left

Chemical Identifiers

CAS 6812-14-2
Molecular Formula C16H8O3
Molecular Weight (g/mol) 248.237
InChI Key AJXNLGUENUIIRW-UHFFFAOYSA-N
PubChem CID 11096934
IUPAC Name naphtho[2,3-f][2]benzofuran-1,3-dione
SMILES C1=CC=C2C=C3C=C4C(=CC3=CC2=C1)C(=O)OC4=O

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 248.24
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Anthracenedicarboxylic Anhydride