missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,7,8,12,13,17,18-Octaethylporphyrin 95.0+%, TCI America™
$156.59 - $1170.62
Chemical Identifiers
| CAS | 2683-82-1 |
|---|---|
| Molecular Formula | C36H46N4 |
| Molecular Weight (g/mol) | 534.792 |
| MDL Number | MFCD00009627 |
| InChI Key | XFIIGRBIXXECHR-UHFFFAOYSA-N |
| Synonym | octaethylporphine, octaethylporphyrin, 2,3,7,8,12,13,17,18-octaethyl-21h,23h-porphine, 2,3,7,8,12,13,17,18-octaethylporphyrin, octaethylpophine, 21h,23h-porphine, 2,3,7,8,12,13,17,18-octaethyl, octaethylporphyrine, 2,3,7,8,12,13,17,18-octaethyl-21h,23h-porphin, acmc-209gte, hciifbhdbocsaf-muzkialcsa-n |
| PubChem CID | 102311 |
| IUPAC Name | 2,3,7,8,12,13,17,18-octaethyl-21,22-dihydroporphyrin |
| SMILES | CCC1=C(C2=CC3=NC(=CC4=NC(=CC5=C(C(=C(N5)C=C1N2)CC)CC)C(=C4CC)CC)C(=C3CC)CC)CC |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
O0234100MG
|
TCI America
O0234100MG |
100 mg |
Each for $156.59
|
|
|||||
|
O02341G
|
TCI America
O02341G |
1 g |
Each for $1,170.62
|
|
|||||
Chemical Identifiers
| 2683-82-1 | |
| 534.792 | |
| XFIIGRBIXXECHR-UHFFFAOYSA-N | |
| 102311 | |
| CCC1=C(C2=CC3=NC(=CC4=NC(=CC5=C(C(=C(N5)C=C1N2)CC)CC)C(=C4CC)CC)C(=C3CC)CC)CC |
| C36H46N4 | |
| MFCD00009627 | |
| octaethylporphine, octaethylporphyrin, 2,3,7,8,12,13,17,18-octaethyl-21h,23h-porphine, 2,3,7,8,12,13,17,18-octaethylporphyrin, octaethylpophine, 21h,23h-porphine, 2,3,7,8,12,13,17,18-octaethyl, octaethylporphyrine, 2,3,7,8,12,13,17,18-octaethyl-21h,23h-porphin, acmc-209gte, hciifbhdbocsaf-muzkialcsa-n | |
| 2,3,7,8,12,13,17,18-octaethyl-21,22-dihydroporphyrin |
Specifications
| 2683-82-1 | |
| MFCD00009627 | |
| octaethylporphine, octaethylporphyrin, 2,3,7,8,12,13,17,18-octaethyl-21h,23h-porphine, 2,3,7,8,12,13,17,18-octaethylporphyrin, octaethylpophine, 21h,23h-porphine, 2,3,7,8,12,13,17,18-octaethyl, octaethylporphyrine, 2,3,7,8,12,13,17,18-octaethyl-21h,23h-porphin, acmc-209gte, hciifbhdbocsaf-muzkialcsa-n | |
| CCC1=C(C2=CC3=NC(=CC4=NC(=CC5=C(C(=C(N5)C=C1N2)CC)CC)C(=C4CC)CC)C(=C3CC)CC)CC | |
| 534.792 | |
| 534.79 | |
| Crystalline Powder |
| C36H46N4 | |
| 100 mg | |
| XFIIGRBIXXECHR-UHFFFAOYSA-N | |
| 2,3,7,8,12,13,17,18-octaethyl-21,22-dihydroporphyrin | |
| 102311 | |
| ≥95.0% (N) | |
| 2,3,7,8,12,13,17,18-Octaethylporphyrin |
Safety and Handling
TSCA : Yes