accessibility menu, dialog, popup

UserName

CAS RN 2683-82-1

H3CNHNNHNCH3CH3CH3H3CH3CH3CH3C

CAS RN 2683-82-1

IUPAC Name: 4,5,9,10,14,15,19,20-octaethyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11,13(22),14,16,18(21),19-undecaene
Synonyms: 4,5,9,10,14,15,19,20-octaethyl-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11,13(22),14,16,18(21),19-undecaene
Molecular Weight (g/mol): 534.79
Molecular Formula: C36H46N4
InChi Key: XFIIGRBIXXECHR-UHFFFAOYSA-N
SMILES: CCC1=C(CC)C2=CC3=NC(=CC4=NC(=CC5=C(CC)C(CC)=C(N5)C=C1N2)C(CC)=C4CC)C(CC)=C3CC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results