missing translation for 'onlineSavingsMsg'
Learn More

2,3,7,8,12,13,17,18-Octaethylporphyrin 95.0+%, TCI America™

Catalog No. O02341G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02341G 1 g
O0234100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02341G Supplier TCI America Supplier No. O02341G
Only null left

Chemical Identifiers

CAS 2683-82-1
Molecular Formula C36H46N4
Molecular Weight (g/mol) 534.792
MDL Number MFCD00009627
InChI Key XFIIGRBIXXECHR-UHFFFAOYSA-N
Synonym octaethylporphine, octaethylporphyrin, 2,3,7,8,12,13,17,18-octaethyl-21h,23h-porphine, 2,3,7,8,12,13,17,18-octaethylporphyrin, octaethylpophine, 21h,23h-porphine, 2,3,7,8,12,13,17,18-octaethyl, octaethylporphyrine, 2,3,7,8,12,13,17,18-octaethyl-21h,23h-porphin, acmc-209gte, hciifbhdbocsaf-muzkialcsa-n
PubChem CID 102311
IUPAC Name 2,3,7,8,12,13,17,18-octaethyl-21,22-dihydroporphyrin
SMILES CCC1=C(C2=CC3=NC(=CC4=NC(=CC5=C(C(=C(N5)C=C1N2)CC)CC)C(=C4CC)CC)C(=C3CC)CC)CC

Specifications

Quantity 1 g
Formula Weight 534.79
Percent Purity ≥95.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,7,8,12,13,17,18-Octaethylporphyrin