missing translation for 'onlineSavingsMsg'
Learn More

2,3,6-Trimethylphenol 98.0+%, TCI America™

Catalog No. p-7136475
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T087725G 25 g
T0877500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T087725G Supplier TCI America Supplier No. T087725G
Only null left

Chemical Identifiers

CAS 2416-94-6
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
MDL Number MFCD00002229
InChI Key QQOMQLYQAXGHSU-UHFFFAOYSA-N
Synonym 3-hydroxypseudocumene, phenol, 2,3,6-trimethyl, 1-hydroxy-2,3,6-trimethylbenzene, 2,3,6-trimethyl-phenol, unii-05wkl2l5lj, 2,3,6-trimethyl phenol, 05wkl2l5lj, dsstox_cid_2187, dsstox_rid_76518, dsstox_gsid_22187
PubChem CID 17016
IUPAC Name 2,3,6-trimethylphenol
SMILES CC1=CC=C(C)C(O)=C1C

Specifications

Melting Point 62°C
Color White-Yellow
Boiling Point 120°C
Quantity 25 g
UN Number 1759
Formula Weight 136.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,6-Trimethylphenol
EINECSNumber (3)-0540&(4)-0057&(9)-0899
TSCA Yes