missing translation for 'onlineSavingsMsg'
Learn More

2,3,6-Trimethoxybenzoic Acid 98.0+%, TCI America™

Catalog No. T15635G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15635G 5 g
T15631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15635G Supplier TCI America Supplier No. T15635G

Chemical Identifiers

CAS 60241-74-9
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.201
MDL Number MFCD00092587
InChI Key NSKSXYUOXAQHCH-UHFFFAOYSA-N
PubChem CID 611400
IUPAC Name 2,3,6-trimethoxybenzoic acid
SMILES COC1=C(C(=C(C=C1)OC)OC)C(=O)O

Specifications

Melting Point 150°C
Color White
Quantity 5 g
Formula Weight 212.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,6-Trimethoxybenzoic Acid