missing translation for 'onlineSavingsMsg'
Learn More

2,3,6-Trifluorophenol 95.0+%, TCI America™

Catalog No. p-7137594
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31291G 1 g
T31295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T31291G Supplier TCI America Supplier No. T31291G
Only null left

Chemical Identifiers

CAS 113798-74-6
Molecular Formula C6H3F3O
Molecular Weight (g/mol) 148.08
MDL Number MFCD00061214
InChI Key QSFGUSFDWCVXNR-UHFFFAOYSA-N
PubChem CID 517988
IUPAC Name 2,3,6-trifluorophenol
SMILES OC1=C(F)C=CC(F)=C1F

Specifications

Color White-Yellow
Quantity 1 g
UN Number 1325
Synonym phenol, 2,3,6-trifluoro, acmc-20aoho, pubchem1504, 2,3,6-trifluoro phenol, phenol,2,3,6-trifluoro, 2,3,6-tris fluoranyl phenol, 2,3,6-trifluorophenol, 2,3,6-trifluorophenol 1g, phenol, 2,3,6-trifluoro-9ci
Formula Weight 148.08
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 2,3,6-Trifluorophenol