missing translation for 'onlineSavingsMsg'
Learn More

2,3,6-Trifluorophenol, 98%

Catalog No. p-7025060
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2037906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2037906 Supplier Thermo Scientific Chemicals Supplier No. B2037906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 113798-74-6 | C6H3F3O | 148.08 g/mol

2,3,6-Trifluorophenol may be employed as model compound to investigate the site-selective functionalization of halogen-bearing phenols by organometallic approach.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,6-Trifluorophenol may be employed as model compound to investigate the site-selective functionalization of halogen-bearing phenols by organometallic approach.

Solubility
Soluble in water (47.267 mg/L est.)

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 113798-74-6
Molecular Formula C6H3F3O
Molecular Weight (g/mol) 148.08
MDL Number MFCD00061214
InChI Key QSFGUSFDWCVXNR-UHFFFAOYSA-N
PubChem CID 517988
IUPAC Name 2,3,6-trifluorophenol
SMILES OC1=C(F)C=CC(F)=C1F

Specifications

Melting Point 31°C to 36°C
Flash Point 63°C (145°F)
Quantity 5 g
UN Number UN3261
Beilstein 4976927
Synonym phenol, 2,3,6-trifluoro, acmc-20aoho, pubchem1504, 2,3,6-trifluoro phenol, phenol,2,3,6-trifluoro, 2,3,6-tris fluoranyl phenol, 2,3,6-trifluorophenol, 2,3,6-trifluorophenol 1g, phenol, 2,3,6-trifluoro-9ci
Solubility Information Soluble in water (47.267mg/L est.)
Formula Weight 148.09
Percent Purity 98%
Chemical Name or Material 2,3,6-Trifluorophenol
Hazard Category H302+H312-H314-H335
Hazard Statement GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only