missing translation for 'onlineSavingsMsg'
Learn More

2,3,6-Trifluorobenzaldehyde 98.0+%, TCI America™

Catalog No. p-7137155
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23181G 1 g
T23185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23181G Supplier TCI America Supplier No. T23181G

Chemical Identifiers

CAS 104451-70-9
Molecular Formula C7H3F3O
Molecular Weight (g/mol) 160.10
MDL Number MFCD00061195
InChI Key XSBAHBVACIKRTG-UHFFFAOYSA-N
Synonym benzaldehyde, 2,3,6-trifluoro, 2,3,6-trifluoro-benzaldehyde, pubchem1467, acmc-1bqm6, 2,3,6-trifluorobenzoaldehyde, ksc494g2t, 2,3,6-trifluoro benzaldehyde, timtec-bb sbb003970, attercop-chm at111497
PubChem CID 517845
IUPAC Name 2,3,6-trifluorobenzaldehyde
SMILES FC1=CC=C(F)C(C=O)=C1F

Specifications

Color Yellow
Boiling Point 159°C
Quantity 1 g
Formula Weight 160.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,6-Trifluorobenzaldehyde