missing translation for 'onlineSavingsMsg'
Learn More

2-(3,6-Dihexyloxyxanthen-9-yl)benzamide 97.0+%, TCI America™

Catalog No. d414225g
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D414225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D414225G Supplier TCI America Supplier No. D414225G

Chemical Identifiers

CAS 1820618-83-4
Molecular Formula C32H39NO4
Molecular Weight (g/mol) 501.667
InChI Key MTYUAGPQJCRKEE-UHFFFAOYSA-N
PubChem CID 56973697
IUPAC Name 2-(3,6-dihexoxy-9H-xanthen-9-yl)benzamide
SMILES CCCCCCOC1=CC2=C(C=C1)C(C3=C(O2)C=C(C=C3)OCCCCCC)C4=CC=CC=C4C(=O)N

Specifications

Melting Point 127°C
Color White-Yellow
Quantity 25 g
Formula Weight 501.67
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-(3,6-Dihexyloxyxanthen-9-yl)benzamide