accessibility menu, dialog, popup

UserName

H3COOOCH3OH2N

CAS RN 1820618-83-4

IUPAC Name: 2-[3,6-bis(hexyloxy)-9H-xanthen-9-yl]benzamide
Synonyms: 2-[3,6-bis(hexyloxy)-9H-xanthen-9-yl]benzamide
Molecular Weight (g/mol): 501.67
Molecular Formula: C32H39NO4
InChi Key: MTYUAGPQJCRKEE-UHFFFAOYSA-N
SMILES: CCCCCCOC1=CC2=C(C=C1)C(C1=CC=CC=C1C(N)=O)C1=C(O2)C=C(OCCCCCC)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results