missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,6,7,10,11-Hexamethoxytriphenylene 95.0+%, TCI America™
$270.29 - $812.73
Chemical Identifiers
| CAS | 808-57-1 |
|---|---|
| Molecular Formula | C24H24O6 |
| Molecular Weight (g/mol) | 408.45 |
| MDL Number | MFCD00075571 |
| InChI Key | TXROZCSFVVIBFI-UHFFFAOYSA-N |
| PubChem CID | 4607363 |
| IUPAC Name | 2,3,6,7,10,11-hexamethoxytriphenylene |
| SMILES | COC1=C(C=C2C(=C1)C3=CC(=C(C=C3C4=CC(=C(C=C24)OC)OC)OC)OC)OC |
Chemical Identifiers
| 808-57-1 | |
| 408.45 | |
| TXROZCSFVVIBFI-UHFFFAOYSA-N | |
| 2,3,6,7,10,11-hexamethoxytriphenylene |
| C24H24O6 | |
| MFCD00075571 | |
| 4607363 | |
| COC1=C(C=C2C(=C1)C3=CC(=C(C=C3C4=CC(=C(C=C24)OC)OC)OC)OC)OC |
Specifications
| 808-57-1 | |
| C24H24O6 | |
| 1 g | |
| COC1=C(C=C2C(=C1)C3=CC(=C(C=C3C4=CC(=C(C=C24)OC)OC)OC)OC)OC | |
| 408.45 | |
| 408.45 | |
| Crystalline Powder |
| Purple | |
| MFCD00075571 | |
| TXROZCSFVVIBFI-UHFFFAOYSA-N | |
| 2,3,6,7,10,11-hexamethoxytriphenylene | |
| 4607363 | |
| ≥95.0% (GC) | |
| 2,3,6,7,10,11-Hexamethoxytriphenylene |
Safety and Handling
EINECSNumber : (4)-1920
TSCA : No