missing translation for 'onlineSavingsMsg'
Learn More

2,3,6,7,10,11-Hexamethoxytriphenylene 95.0+%, TCI America™

Catalog No. H08981G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08981G 1 g
H08985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08981G Supplier TCI America Supplier No. H08981G
Only null left

Chemical Identifiers

CAS 808-57-1
Molecular Formula C24H24O6
Molecular Weight (g/mol) 408.45
MDL Number MFCD00075571
InChI Key TXROZCSFVVIBFI-UHFFFAOYSA-N
PubChem CID 4607363
IUPAC Name 2,3,6,7,10,11-hexamethoxytriphenylene
SMILES COC1=C(C=C2C(=C1)C3=CC(=C(C=C3C4=CC(=C(C=C24)OC)OC)OC)OC)OC

Specifications

Color Purple
Quantity 1 g
Formula Weight 408.45
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,6,7,10,11-Hexamethoxytriphenylene
EINECSNumber (4)-1920
TSCA No