missing translation for 'onlineSavingsMsg'
Learn More

2,3,6,7,10,11-Hexakis(hexyloxy)triphenylene 98.0+%, TCI America™

Catalog No. p-7131049
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H1449200MG 200 mg
H14491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H1449200MG Supplier TCI America Supplier No. H1449200MG
Only null left

Chemical Identifiers

CAS 70351-86-9
Molecular Formula C54H84O6
Molecular Weight (g/mol) 829.26
MDL Number MFCD16250223
InChI Key SQYCPYUKCAKHRN-UHFFFAOYSA-N
PubChem CID 10941839
IUPAC Name 2,3,6,7,10,11-hexakis(hexyloxy)triphenylene
SMILES CCCCCCOC1=C(OCCCCCC)C=C2C(=C1)C1=CC(OCCCCCC)=C(OCCCCCC)C=C1C1=CC(OCCCCCC)=C(OCCCCCC)C=C21

Specifications

Color White-Yellow
Quantity 200 mg
Formula Weight 829.26
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,6,7,10,11-Hexakis(hexyloxy)triphenylene