missing translation for 'onlineSavingsMsg'
Learn More

2,3,6,7,10,11-Hexahydroxytriphenylene Hydrate 95.0+%, TCI America™

Catalog No. H09075G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09075G 5 g
H09071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09075G Supplier TCI America Supplier No. H09075G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4877-80-9
Molecular Formula C18H12O6
Molecular Weight (g/mol) 324.288
MDL Number MFCD01321170
InChI Key QMLILIIMKSKLES-UHFFFAOYSA-N
Synonym 2,3,6,7,10,11-hexahydroxytriphenylene, triphenylene-2,3,6,7,10,11-hexaol, 2,3,6,7,10,11-triphenylenehexol, acmc-1ak9f, 2,3,6,7,10,11-hexa-hydroxytriphenylene
PubChem CID 11088610
IUPAC Name triphenylene-2,3,6,7,10,11-hexol
SMILES C1=C2C3=CC(=C(C=C3C4=CC(=C(C=C4C2=CC(=C1O)O)O)O)O)O

Specifications

Quantity 5 g
Formula Weight 324.29
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,6,7,10,11-Hexahydroxytriphenylene Hydrate
EINECSNumber (4)-1890
TSCA No