missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3,5-Triphenyltetrazolium Chloride, Reagents
Specifications
Specifications
| Color | White to Yellow |
| Packaging | Glass Bottle |
| Quantity | 100 g |
| Specific Gravity | 1 |
| Grade | Reagent |
| Physical Form | Solid |
| Chemical Name or Material | 2,3,5-Triphenyltetrazolium Chloride |